Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCATTCTTCTCATACATCATTTGT[C/G]CATTAACAGGAAAAAAGTCTCACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf40 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf40 - chromosome 11 open reading frame 40 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR52I1 - olfactory receptor family 52 subfamily I member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR52I2 - olfactory receptor family 52 subfamily I member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005170.2 | 72 | Missense Mutation | TCC,TGC | S,C 17 | NP_001005170.1 |