Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCTTCCTGCTGGCCGTGATGGCC[C/T]ATGACCGCTTTGTGGCCATCTGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR5D18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR5D18 - olfactory receptor family 5 subfamily D member 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR5L1 - olfactory receptor family 5 subfamily L member 1 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004738.1 | 358 | Missense Mutation | CAT,TAT | H,Y 120 | NP_001004738.1 |