Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAAACTCACATCATAAGGTTCCG[A/G]TGCATTGTCTAATCGCAGAACTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610110 MIM: 608566 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANO3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANO3 - anoctamin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313726.1 | 1219 | Intron | NP_001300655.1 | |||
NM_001313727.1 | 1219 | Intron | NP_001300656.1 | |||
NM_031418.3 | 1219 | Intron | NP_113606.2 | |||
XM_011520282.2 | 1219 | Intron | XP_011518584.1 | |||
XM_017018118.1 | 1219 | Intron | XP_016873607.1 | |||
XM_017018119.1 | 1219 | Intron | XP_016873608.1 |
MUC15 - mucin 15, cell surface associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135091.1 | 1219 | Missense Mutation | CCG,TCG | P,S 316 | NP_001128563.1 | |
NM_001135092.1 | 1219 | Missense Mutation | CCG,TCG | P,S 266 | NP_001128564.1 | |
NM_145650.3 | 1219 | Missense Mutation | CCG,TCG | P,S 289 | NP_663625.2 | |
XM_017017235.1 | 1219 | Missense Mutation | CCG,TCG | P,S 405 | XP_016872724.1 | |
XM_017017236.1 | 1219 | Missense Mutation | CCG,TCG | P,S 355 | XP_016872725.1 |