Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGCAAATAAAAAGGATACACAT[A/G]ACGGGCCACCAAAAGAAATTAAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC83 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC83 - coiled-coil domain containing 83 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286159.1 | 552 | Missense Mutation | AAC,GAC | N,D 14 | NP_001273088.1 | |
NM_173556.4 | 552 | Missense Mutation | AAC,GAC | N,D 14 | NP_775827.2 | |
XM_011544839.2 | 552 | Missense Mutation | AAC,GAC | N,D 14 | XP_011543141.1 | |
XM_011544840.2 | 552 | Missense Mutation | AAC,GAC | N,D 14 | XP_011543142.1 | |
XM_011544841.1 | 552 | Missense Mutation | AAC,GAC | N,D 14 | XP_011543143.1 | |
XM_011544842.1 | 552 | Missense Mutation | AAC,GAC | N,D 14 | XP_011543144.1 | |
XM_011544843.1 | 552 | Missense Mutation | AAC,GAC | N,D 14 | XP_011543145.1 | |
XM_011544844.2 | 552 | Intron | XP_011543146.1 | |||
XM_017017354.1 | 552 | Missense Mutation | AAC,GAC | N,D 14 | XP_016872843.1 | |
XM_017017355.1 | 552 | Missense Mutation | AAC,GAC | N,D 14 | XP_016872844.1 |