Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAGTCGGTATAGGGTGGGTAGT[A/G]GATCTCCTTTAATTTCTCCAGGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601917 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH3B2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH3B2 - aldehyde dehydrogenase 3 family member B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000695.3 | 1364 | Missense Mutation | CAC,TAC | H,Y 361 | NP_000686.2 | |
NM_001031615.1 | 1364 | Missense Mutation | CAC,TAC | H,Y 361 | NP_001026786.1 |