Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCTGGGCAGAGGCTGTGGAACTC[A/G]GCTGCAGGGGCAGGGCGGCCGTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602090 MIM: 607982 | ||||||||||||||||||||
Literature Links: |
LTBP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LTBP3 - latent transforming growth factor beta binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130144.2 | 3346 | Silent Mutation | GCC,GCT | A,A 965 | NP_001123616.1 | |
NM_001164266.1 | 3346 | Silent Mutation | GCC,GCT | A,A 848 | NP_001157738.1 | |
NM_021070.4 | 3346 | Silent Mutation | GCC,GCT | A,A 965 | NP_066548.2 | |
XM_011545032.2 | 3346 | Silent Mutation | GCC,GCT | A,A 974 | XP_011543334.1 | |
XM_011545033.2 | 3346 | Silent Mutation | GCC,GCT | A,A 974 | XP_011543335.1 | |
XM_017017737.1 | 3346 | Intron | XP_016873226.1 |
SCYL1 - SCY1 like pseudokinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |