Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTTGTCATCCCGCCAAACAACG[A/C]CAGTGGCCTCTGCCCACCTCCGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MS4A15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MS4A15 - membrane spanning 4-domains A15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098835.1 | 276 | Missense Mutation | GAC,GCC | D,A 19 | NP_001092305.1 | |
NM_001278242.1 | 276 | Missense Mutation | GAC,GCC | D,A 19 | NP_001265171.1 | |
NM_152717.2 | 276 | Intron | NP_689930.1 | |||
XM_006718459.3 | 276 | Intron | XP_006718522.1 | |||
XM_011544812.2 | 276 | Missense Mutation | GAC,GCC | D,A 19 | XP_011543114.1 | |
XM_011544814.1 | 276 | UTR 5 | XP_011543116.1 |