Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCCTGAGAAATAAAGATGTGAA[A/G]GAAGCTCTCAGAAAAGTGATGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR5D18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR5D18 - olfactory receptor family 5 subfamily D member 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR5L1 - olfactory receptor family 5 subfamily L member 1 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004738.1 | 894 | Missense Mutation | AAA,AAG | K,K 298 | NP_001004738.1 |