Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616097 | ||||||||||||||||||||
Literature Links: |
C11orf98 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C11orf98 - chromosome 11 open reading frame 98 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286086.1 | 820 | Intron | NP_001273015.1 |
LBHD1 - LBH domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024099.3 | 820 | Missense Mutation | AGT,GGT | S,G 202 | NP_077004.2 |
METTL12 - methyltransferase like 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001043229.1 | 820 | Intron | NP_001036694.1 | |||
XM_005274232.4 | 820 | Intron | XP_005274289.1 |
SNORA57 - small nucleolar RNA, H/ACA box 57 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UQCC3 - ubiquinol-cytochrome c reductase complex assembly factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |