Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTCTTCTTCTCAAGGTCGTAAT[C/T]GAACCAGGCTCCGGTCTGCTCATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612834 MIM: 275360 | ||||||||||||||||||||
Literature Links: |
PHLDB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PHLDB1 - pleckstrin homology like domain family B member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TREH - trehalase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301065.1 | 1246 | Missense Mutation | AAT,GAT | N,D 370 | NP_001287994.1 | |
NM_007180.2 | 1246 | Missense Mutation | AAT,GAT | N,D 401 | NP_009111.2 |