Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTCTAATAAGTTCTGCTAGGTC[G/T]TCTGTATTCTGAGATTTTTCTTCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603652 | ||||||||||||||||||||
Literature Links: |
TRPC6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRPC6 - transient receptor potential cation channel subfamily C member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004621.5 | 2648 | Missense Mutation | GAA,GAC | E,D 907 | NP_004612.2 | |
XM_011542968.2 | 2648 | Missense Mutation | GAA,GAC | E,D 852 | XP_011541270.1 | |
XM_017018221.1 | 2648 | Missense Mutation | GAA,GAC | E,D 791 | XP_016873710.1 | |
XM_017018222.1 | 2648 | Missense Mutation | GAA,GAC | E,D 821 | XP_016873711.1 |