Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAATGGGCTTCAGGGCCCGCTCC[A/C]AGGCATTCACCAATGAAGGCAGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603864 | ||||||||||||||||||||
Literature Links: |
CCDC87 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC87 - coiled-coil domain containing 87 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018219.2 | 2230 | Missense Mutation | TGG,TTG | W,L 721 | NP_060689.2 |
CCS - copper chaperone for superoxide dismutase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |