Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATTTTTCTTCAAGGAGTTCATA[A/G]CGGAGACTTGAGATGTCCTGCTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603652 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRPC6 - transient receptor potential cation channel subfamily C member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004621.5 | 2612 | Silent Mutation | CGC,CGT | R,R 895 | NP_004612.2 | |
XM_011542968.2 | 2612 | Silent Mutation | CGC,CGT | R,R 840 | XP_011541270.1 | |
XM_017018221.1 | 2612 | Silent Mutation | CGC,CGT | R,R 779 | XP_016873710.1 | |
XM_017018222.1 | 2612 | Silent Mutation | CGC,CGT | R,R 809 | XP_016873711.1 |