Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCCTCATGTTCTTCAACACCTT[C/T]GTCTTCCAGGCTGGCCTGGTCAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614459 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYB561A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYB561A3 - cytochrome b561 family member A3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM138 - transmembrane protein 138 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016464.4 | 391 | Silent Mutation | TTC,TTT | F,F 64 | NP_057548.1 | |
XM_006718585.3 | 391 | Silent Mutation | TTC,TTT | F,F 64 | XP_006718648.1 | |
XM_006718586.2 | 391 | Silent Mutation | TTC,TTT | F,F 64 | XP_006718649.1 | |
XM_006718588.3 | 391 | Silent Mutation | TTC,TTT | F,F 6 | XP_006718651.1 | |
XM_011545098.2 | 391 | Silent Mutation | TTC,TTT | F,F 64 | XP_011543400.1 | |
XM_011545099.1 | 391 | Silent Mutation | TTC,TTT | F,F 64 | XP_011543401.1 | |
XM_011545100.1 | 391 | Silent Mutation | TTC,TTT | F,F 64 | XP_011543402.1 | |
XM_017017917.1 | 391 | Silent Mutation | TTC,TTT | F,F 64 | XP_016873406.1 |