Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATCCACTAGCATCTTACCTTGC[A/G]GAAAATTTCCTCCACATCACAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147678 MIM: 602665 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASP1 - caspase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001223.4 | 1092 | Missense Mutation | CGC,TGC | R,C 350 | NP_001214.1 | |
NM_001257118.2 | 1092 | Missense Mutation | CGC,TGC | R,C 371 | NP_001244047.1 | |
NM_001257119.2 | 1092 | Missense Mutation | CGC,TGC | R,C 350 | NP_001244048.1 | |
NM_033292.3 | 1092 | Missense Mutation | CGC,TGC | R,C 371 | NP_150634.1 | |
NM_033293.3 | 1092 | Missense Mutation | CGC,TGC | R,C 278 | NP_150635.1 | |
NM_033294.3 | 1092 | Missense Mutation | CGC,TGC | R,C 230 | NP_150636.1 | |
NM_033295.3 | 1092 | Missense Mutation | CGC,TGC | R,C 55 | NP_150637.1 | |
XM_017018393.1 | 1092 | Missense Mutation | CGC,TGC | R,C 368 | XP_016873882.1 | |
XM_017018394.1 | 1092 | Missense Mutation | CGC,TGC | R,C 368 | XP_016873883.1 | |
XM_017018395.1 | 1092 | Missense Mutation | CGC,TGC | R,C 347 | XP_016873884.1 | |
XM_017018396.1 | 1092 | Missense Mutation | CGC,TGC | R,C 341 | XP_016873885.1 |
CASP5 - caspase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |