Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608213 MIM: 605247 MIM: 609302 | ||||||||||||||||||||
Literature Links: |
CEND1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEND1 - cell cycle exit and neuronal differentiation 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC171391 - uncharacterized LOC171391 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PANO1 - proapoptotic nucleolar protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIDD1 - p53-induced death domain protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A22 - solute carrier family 25 member 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191060.1 | 1110 | Missense Mutation | AAG,GAG | K,E 262 | NP_001177989.1 | |
NM_001191061.1 | 1110 | Missense Mutation | AAG,GAG | K,E 262 | NP_001177990.1 | |
NM_024698.5 | 1110 | Missense Mutation | AAG,GAG | K,E 262 | NP_078974.1 | |
XM_011520369.1 | 1110 | Missense Mutation | AAG,GAG | K,E 262 | XP_011518671.1 | |
XM_011520370.1 | 1110 | Missense Mutation | AAG,GAG | K,E 262 | XP_011518672.1 | |
XM_011520371.1 | 1110 | Missense Mutation | AAG,GAG | K,E 262 | XP_011518673.1 |