Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTAAGGCAGGGTGCCATACTAAG[C/G]AGCTTGCCTCCAGGAATTGCTCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604872 | ||||||||||||||||||||
Literature Links: |
PCSK7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCSK7 - proprotein convertase subtilisin/kexin type 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF214 - ring finger protein 214 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077239.1 | 535 | Missense Mutation | CAG,GAG | Q,E 143 | NP_001070707.1 | |
NM_001278249.1 | 535 | Intron | NP_001265178.1 | |||
NM_207343.3 | 535 | Missense Mutation | CAG,GAG | Q,E 143 | NP_997226.2 |