Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCAGCAGGCCCCACACGTGCTCCT[C/T]GGGCACTGCCAGGCCCAGCTCCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604648 | ||||||||||||||||||||
Literature Links: |
NUDT8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUDT8 - nudix hydrolase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243750.1 | 312 | Missense Mutation | NP_001230679.1 | |||
NM_181843.2 | 312 | Missense Mutation | NP_862826.1 | |||
XM_017017474.1 | 312 | Nonsense Mutation | XP_016872963.1 |
TBX10 - T-box 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |