Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGTGGCTCAATGGCTCTCTACTT[C/T]GAATTGGACCTGGGAAATTCGAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611740 MIM: 605791 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCO2 - beta-carotene oxygenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037290.2 | 373 | Silent Mutation | CGA,TGA | R,* 52 | NP_001032367.2 | |
NM_001256397.1 | 373 | Silent Mutation | CGA,TGA | R,* 52 | NP_001243326.1 | |
NM_001256398.1 | 373 | Silent Mutation | CGA,TGA | R,* 86 | NP_001243327.1 | |
NM_001256400.1 | 373 | Intron | NP_001243329.1 | |||
NM_031938.5 | 373 | Silent Mutation | CGA,TGA | R,* 86 | NP_114144.4 |
TEX12 - testis expressed 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |