Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGAGCGGCAGCAAGTCGTCCGG[A/G]CCGCCACCGCCGTCGGGTTCCTCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601178 | ||||||||||||||||||||
Literature Links: |
CAPRIN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAPRIN1 - cell cycle associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005898.4 | 237 | Silent Mutation | GGA,GGG | G,G 16 | NP_005889.3 | |
NM_203364.2 | 237 | Silent Mutation | GGA,GGG | G,G 16 | NP_976240.1 | |
XM_017017749.1 | 237 | Silent Mutation | GGA,GGG | G,G 16 | XP_016873238.1 |