Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACGAGGACTTCAACGTCCAGCTAC[A/G]CCGAGGCCTAGTGGGCTCCAGGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600770 | ||||||||||||||||||||
Literature Links: |
MUC5B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MUC5B - mucin 5B, oligomeric mucus/gel-forming | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002458.2 | 415 | Missense Mutation | CAC,CGC | H,R 119 | NP_002449.2 |