Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCAAGCCCATTCCTGCAACGCTT[A/G]CAGCGATGATGACATCTCTGACCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613866 MIM: 613867 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HRASLS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HRASLS2 - HRAS like suppressor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLA2G16 - phospholipase A2 group XVI | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128203.1 | 735 | Missense Mutation | GCA,GTA | A,V 137 | NP_001121675.1 | |
NM_007069.3 | 735 | Missense Mutation | GCA,GTA | A,V 137 | NP_009000.2 | |
XM_006718426.1 | 735 | Missense Mutation | GCA,GTA | A,V 137 | XP_006718489.1 | |
XM_011544741.1 | 735 | Missense Mutation | GCA,GTA | A,V 152 | XP_011543043.1 |