Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAGAGGACTAACCGGGGACTTG[A/T]CCCTGGCGCCGGGCCGCCCGTTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613991 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC42BPG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDC42BPG - CDC42 binding protein kinase gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017525.2 | 4528 | Missense Mutation | GAC,GTC | D,V 1460 | NP_059995.2 | |
XM_011545155.2 | 4528 | Missense Mutation | GAC,GTC | D,V 1478 | XP_011543457.1 | |
XM_011545156.2 | 4528 | Missense Mutation | GAC,GTC | D,V 1471 | XP_011543458.1 | |
XM_011545157.2 | 4528 | Silent Mutation | GGA,GGT | G,G 1445 | XP_011543459.1 | |
XM_011545158.2 | 4528 | Silent Mutation | GGA,GGT | G,G 1445 | XP_011543460.1 | |
XM_011545159.2 | 4528 | Missense Mutation | GAC,GTC | D,V 1478 | XP_011543461.1 | |
XM_011545160.2 | 4528 | Missense Mutation | GAC,GTC | D,V 1380 | XP_011543462.1 | |
XM_011545161.2 | 4528 | Intron | XP_011543463.1 | |||
XM_017017996.1 | 4528 | Missense Mutation | GAC,GTC | D,V 1477 | XP_016873485.1 | |
XM_017017997.1 | 4528 | Intron | XP_016873486.1 | |||
XM_017017998.1 | 4528 | Intron | XP_016873487.1 | |||
XM_017017999.1 | 4528 | Missense Mutation | GAC,GTC | D,V 774 | XP_016873488.1 |