Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAATGGGCAGTGAGAGGTCCCAGA[A/C]GGGGTCGAAGACCGTAGAACAGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604725 | ||||||||||||||||||||
Literature Links: |
USP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
USP2 - ubiquitin specific peptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243759.1 | 568 | Missense Mutation | GTC,TTC | V,F 195 | NP_001230688.1 | |
NM_004205.4 | 568 | Missense Mutation | GTC,TTC | V,F 438 | NP_004196.4 | |
NM_171997.2 | 568 | Missense Mutation | GTC,TTC | V,F 229 | NP_741994.1 | |
XM_005271721.4 | 568 | Missense Mutation | GTC,TTC | V,F 438 | XP_005271778.1 | |
XM_005271722.2 | 568 | Missense Mutation | GTC,TTC | V,F 438 | XP_005271779.1 | |
XM_017018539.1 | 568 | Missense Mutation | GTC,TTC | V,F 185 | XP_016874028.1 |