Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTCCTCTGCCTCCCGGCGGGTC[G/T]TGGGTGTGTAGAAGAAGCCTCGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176730 MIM: 191290 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INS - insulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000207.2 | 217 | Missense Mutation | AAG,ACG | K,T 53 | NP_000198.1 | |
NM_001185097.1 | 217 | Missense Mutation | AAG,ACG | K,T 53 | NP_001172026.1 | |
NM_001185098.1 | 217 | Missense Mutation | AAG,ACG | K,T 53 | NP_001172027.1 | |
NM_001291897.1 | 217 | Missense Mutation | AAG,ACG | K,T 53 | NP_001278826.1 |
INS-IGF2 - INS-IGF2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042376.2 | 217 | Missense Mutation | AAG,ACG | K,T 53 | NP_001035835.1 |
TH - tyrosine hydroxylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |