Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCAATGGAGCCCCTCTCCTAGCC[G/T]CCTCCATGGGGACTGGGGCTCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600526 MIM: 616019 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MARK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MARK2 - microtubule affinity regulating kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RCOR2 - REST corepressor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173587.3 | 1593 | Missense Mutation | GAG,GCG | E,A 402 | NP_775858.2 | |
XM_005273932.3 | 1593 | Intron | XP_005273989.1 |