Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGGGAAGGTGAGGAGGCTAGGA[C/T]GTGGGACTTCCTGACAGCAGTGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614757 MIM: 609650 MIM: 617032 | ||||||||||||||||||||
Literature Links: |
IFITM5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IFITM5 - interferon induced transmembrane protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NLRP6 - NLR family pyrin domain containing 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PGGHG - protein-glucosylgalactosylhydroxylysine glucosidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025092.4 | 1742 | Missense Mutation | ACG,ATG | T,M 197 | NP_079368.3 | |
XM_011520383.2 | 1742 | Missense Mutation | ACG,ATG | T,M 197 | XP_011518685.1 | |
XM_011520384.2 | 1742 | Missense Mutation | ACG,ATG | T,M 197 | XP_011518686.1 | |
XM_011520385.2 | 1742 | Missense Mutation | ACG,ATG | T,M 197 | XP_011518687.1 | |
XM_011520386.2 | 1742 | Missense Mutation | ACG,ATG | T,M 197 | XP_011518688.1 | |
XM_011520387.2 | 1742 | Missense Mutation | ACG,ATG | T,M 197 | XP_011518689.1 | |
XM_017018355.1 | 1742 | Missense Mutation | ACG,ATG | T,M 197 | XP_016873844.1 |