Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCAGTATTAGCGTGGCCTTGACC[A/T]TCCCATCTGCCATGATGGCCTCCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613277 | ||||||||||||||||||||
Literature Links: |
CPSF7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPSF7 - cleavage and polyadenylation specific factor 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM216 - transmembrane protein 216 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173990.2 | 1248 | Missense Mutation | ATC,TTC | I,F 97 | NP_001167461.1 | |
NM_001173991.2 | 1248 | Missense Mutation | ATC,TTC | I,F 97 | NP_001167462.1 | |
NM_016499.5 | 1248 | Missense Mutation | ATC,TTC | I,F 36 | NP_057583.2 | |
XM_005274039.3 | 1248 | Missense Mutation | ATC,TTC | I,F 36 | XP_005274096.1 | |
XM_017017882.1 | 1248 | Missense Mutation | ATC,TTC | I,F 36 | XP_016873371.1 |