Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGCCGCGCCGGCTTCTCAGCAC[C/T]CCGCAACCGGCACCGGCGCTGTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601178 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPRIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAPRIN1 - cell cycle associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005898.4 | 313 | Missense Mutation | CCC,TCC | P,S 42 | NP_005889.3 | |
NM_203364.2 | 313 | Missense Mutation | CCC,TCC | P,S 42 | NP_976240.1 | |
XM_017017749.1 | 313 | Missense Mutation | CCC,TCC | P,S 42 | XP_016873238.1 |