Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTGTCTTTATTCCTGGCGGGAGA[A/C]CGCACATTCCACTGCTGAGGCTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606871 MIM: 609276 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JAM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JAM3 - junctional adhesion molecule 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCAPD3 - non-SMC condensin II complex subunit D3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015261.2 | 5025 | Silent Mutation | CGG,CGT | R,R 1473 | NP_056076.1 | |
XM_011542719.2 | 5025 | Silent Mutation | CGG,CGT | R,R 1335 | XP_011541021.1 |