Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGTGTCTGGGACAGTCGTACCA[A/G]CTCTGCCTTCTGTACCAGCCTGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606374 MIM: 104160 MIM: 180721 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B3GAT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B3GAT3 - beta-1,3-glucuronyltransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288721.1 | 505 | Silent Mutation | CTG,TTG | L,L 86 | NP_001275650.1 | |
NM_001288722.1 | 505 | Silent Mutation | CTG,TTG | L,L 93 | NP_001275651.1 | |
NM_001288723.1 | 505 | Silent Mutation | CTG,TTG | L,L 93 | NP_001275652.1 | |
NM_012200.3 | 505 | Silent Mutation | CTG,TTG | L,L 93 | NP_036332.2 |
EML3 - echinoderm microtubule associated protein like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GANAB - glucosidase II alpha subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ROM1 - retinal outer segment membrane protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |