Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCACGTGGGAGCCCTGGCAGGTG[A/C]CTTGGCGGTCACTTCGCACTGTCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609651 MIM: 601644 | ||||||||||||||||||||
Literature Links: |
GPHA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPHA2 - glycoprotein hormone alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130769.3 | 937 | Missense Mutation | GGC,GTC | G,V 46 | NP_570125.1 | |
XM_011544774.2 | 937 | Missense Mutation | GGC,GTC | G,V 79 | XP_011543076.1 | |
XM_011544775.2 | 937 | Missense Mutation | GGC,GTC | G,V 46 | XP_011543077.1 | |
XM_011544776.2 | 937 | Missense Mutation | GGC,GTC | G,V 46 | XP_011543078.1 |
MAJIN - membrane anchored junction protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP2R5B - protein phosphatase 2 regulatory subunit B'beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006244.3 | 937 | Intron | NP_006235.1 | |||
XM_011545132.2 | 937 | Intron | XP_011543434.1 | |||
XM_011545133.1 | 937 | Intron | XP_011543435.1 | |||
XM_011545134.2 | 937 | Intron | XP_011543436.1 |