Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATGGCGGCAGTGGTGGAGGTGG[G/A]GGTTGGAGGTGGTGCTGCTGGGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC100133315 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100133315 - transient receptor potential cation channel, subfamily C, member 2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF121 - ring finger protein 121 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300926.1 | 363 | UTR 5 | NP_001287855.1 | |||
NM_018320.4 | 363 | Missense Mutation | GAG,GGG | E,G 8 | NP_060790.2 | |
XM_006718629.1 | 363 | Missense Mutation | GAG,GGG | E,G 8 | XP_006718692.1 | |
XM_006718630.1 | 363 | UTR 5 | XP_006718693.1 | |||
XM_006718632.1 | 363 | UTR 5 | XP_006718695.1 | |||
XM_011545150.1 | 363 | UTR 5 | XP_011543452.1 |