Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACATATTGCAAGGCTCTGACTC[C/T]GCTTTCTGTGTTCAGCTTCCCCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609191 | ||||||||||||||||||||
Literature Links: |
AKIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKIP1 - A-kinase interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C11orf16 - chromosome 11 open reading frame 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020643.2 | 1943 | Missense Mutation | CAG,CGG | Q,R 453 | NP_065694.2 | |
XM_017018013.1 | 1943 | Missense Mutation | CAG,CGG | Q,R 453 | XP_016873502.1 |