Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCGAAGCTATGAGTGGAGCTGCC[A/G]CCTGGTCACCTTCACCCATGGAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604763 MIM: 607394 | ||||||||||||||||||||
Literature Links: |
ARHGEF12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGEF12 - Rho guanine nucleotide exchange factor 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POU2F3 - POU class 2 homeobox 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM136 - transmembrane protein 136 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198670.1 | 340 | Missense Mutation | CAC,CGC | H,R 59 | NP_001185599.1 | |
NM_001198671.1 | 340 | Missense Mutation | CAC,CGC | H,R 37 | NP_001185600.1 | |
NM_001198672.1 | 340 | UTR 5 | NP_001185601.1 | |||
NM_001198673.1 | 340 | UTR 5 | NP_001185602.1 | |||
NM_001198674.1 | 340 | Missense Mutation | CAC,CGC | H,R 37 | NP_001185603.1 | |
NM_001198675.1 | 340 | Intron | NP_001185604.1 | |||
NM_174926.2 | 340 | Missense Mutation | CAC,CGC | H,R 59 | NP_777586.1 | |
XM_017017327.1 | 340 | Missense Mutation | CAC,CGC | H,R 59 | XP_016872816.1 |