Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGAAGCTGCAGGTATCCAGCCA[C/G]CGCACTGGGCTGTACGGACGGCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600502 MIM: 611834 | ||||||||||||||||||||
Literature Links: |
IGHMBP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGHMBP2 - immunoglobulin mu binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002180.2 | 267 | Missense Mutation | CAC,CAG | H,Q 55 | NP_002171.2 | |
XM_005273974.2 | 267 | UTR 5 | XP_005274031.1 | |||
XM_005273975.3 | 267 | Intron | XP_005274032.1 | |||
XM_005273976.1 | 267 | Missense Mutation | CAC,CAG | H,Q 55 | XP_005274033.1 | |
XM_011544994.1 | 267 | Intron | XP_011543296.1 | |||
XM_017017669.1 | 267 | UTR 5 | XP_016873158.1 | |||
XM_017017670.1 | 267 | Intron | XP_016873159.1 | |||
XM_017017671.1 | 267 | Missense Mutation | CAC,CAG | H,Q 55 | XP_016873160.1 |
MRPL21 - mitochondrial ribosomal protein L21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |