Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACCACCCCAGAGCGCAGTGTGT[C/T]GTTCTTGAAGCCAGGGCTCCTCACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PAMR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAMR1 - peptidase domain containing associated with muscle regeneration 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001991.2 | 2340 | Missense Mutation | AAC,GAC | N,D 615 | NP_001001991.1 | |
NM_001282675.1 | 2340 | Missense Mutation | AAC,GAC | N,D 575 | NP_001269604.1 | |
NM_001282676.1 | 2340 | Missense Mutation | AAC,GAC | N,D 504 | NP_001269605.1 | |
NM_015430.3 | 2340 | Missense Mutation | AAC,GAC | N,D 632 | NP_056245.2 |