Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAAGTCTTCTGAGGCAGTGGGCT[C/T]GACGCCAGTGGCCTCCTGTGAAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605601 MIM: 606814 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRG2 - proteoglycan 2, pro eosinophil major basic protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243245.2 | 813 | Missense Mutation | NP_001230174.1 | |||
NM_001302926.1 | 813 | Missense Mutation | NP_001289855.1 | |||
NM_001302927.1 | 813 | Missense Mutation | NP_001289856.1 | |||
NM_002728.5 | 813 | Missense Mutation | NP_002719.3 |
PRG3 - proteoglycan 3, pro eosinophil major basic protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |