Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTATGTTCCTCCAGAACCGCTCA[A/G]TAAAATTGGTTCTAAGAAAAAGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615680 MIM: 147678 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARD16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CARD16 - caspase recruitment domain family member 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017534.1 | 572 | Missense Mutation | ACT,ATT | T,I 161 | NP_001017534.1 | |
NM_052889.2 | 572 | UTR 3 | NP_443121.1 | |||
XM_011542583.2 | 572 | Missense Mutation | ACT,ATT | T,I 229 | XP_011540885.1 |
CASP1 - caspase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |