Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGGCCGACTGTGTGCCACAGTC[G/T]GACCAGTAGCCCTCAGGTCTTTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612324 MIM: 606666 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC34 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC34 - coiled-coil domain containing 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LGR4 - leucine rich repeat containing G protein-coupled receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018490.2 | 3176 | Silent Mutation | NP_060960.2 |