Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTCCTCGGACATGATCAGCCCA[C/T]CGCTGGGGGACTTCCGCCACACCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606132 | ||||||||||||||||||||
Literature Links: |
CDC42EP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC42EP2 - CDC42 effector protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006779.3 | 700 | Missense Mutation | CCG,TCG | P,S 33 | NP_006770.1 | |
XM_017017094.1 | 700 | Missense Mutation | CCG,TCG | P,S 33 | XP_016872583.1 |