Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGAACTCTGTAGGTCTTCACCAC[A/G]GCTCAGGAGGATGAGGAGCAGTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614660 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PATL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PATL1 - PAT1 homolog 1, processing body mRNA decay factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152716.2 | 3348 | Missense Mutation | NP_689929.2 | |||
XM_005273833.4 | 3348 | Missense Mutation | XP_005273890.1 | |||
XM_005273834.1 | 3348 | Missense Mutation | XP_005273891.1 | |||
XM_011544811.2 | 3348 | Missense Mutation | XP_011543113.1 | |||
XM_017017339.1 | 3348 | Missense Mutation | XP_016872828.1 |