Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCCTTGCGGCCCCCCAGCCGCC[A/G]GGCCTGTCTGTCCATCACACTAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600526 MIM: 616019 | ||||||||||||||||||||
Literature Links: |
MARK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MARK2 - microtubule affinity regulating kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RCOR2 - REST corepressor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173587.3 | 959 | Missense Mutation | CGG,TGG | R,W 191 | NP_775858.2 | |
XM_005273932.3 | 959 | Intron | XP_005273989.1 |