Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGACCCCATGGACCAGCCAGAGGC[C/T]CCCTGCTCCAGGTGAGTGCTGGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609650 MIM: 617032 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLRP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NLRP6 - NLR family pyrin domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276700.1 | 223 | Silent Mutation | GCC,GCT | A,A 6 | NP_001263629.1 | |
NM_138329.2 | 223 | Silent Mutation | GCC,GCT | A,A 6 | NP_612202.2 | |
XM_011519931.1 | 223 | Silent Mutation | GCC,GCT | A,A 6 | XP_011518233.1 | |
XM_017017252.1 | 223 | Silent Mutation | GCC,GCT | A,A 6 | XP_016872741.1 | |
XM_017017253.1 | 223 | Intron | XP_016872742.1 |
PGGHG - protein-glucosylgalactosylhydroxylysine glucosidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |