Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGCCTATGTGAAAAGCTGGTCA[A/G]ATTTCGCGAAGGTGAACGCCCGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609641 | ||||||||||||||||||||
Literature Links: |
EIF3M PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EIF3M - eukaryotic translation initiation factor 3 subunit M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307929.1 | 420 | Intron | NP_001294858.1 | |||
NM_006360.5 | 420 | Missense Mutation | AAA,AGA | K,R 92 | NP_006351.2 | |
XM_011519840.2 | 420 | Missense Mutation | AAA,AGA | K,R 29 | XP_011518142.1 |