Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTTGGGGGTCAGGAAGATCTCA[A/T]CCCGTGGGGAGGGAGGTGATGAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608630 MIM: 607528 | ||||||||||||||||||||
Literature Links: |
ROBO3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ROBO3 - roundabout guidance receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ROBO4 - roundabout guidance receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301088.1 | 2962 | Missense Mutation | GAT,GTT | D,V 796 | NP_001288017.1 | |
NM_019055.5 | 2962 | Missense Mutation | GAT,GTT | D,V 941 | NP_061928.4 | |
XM_006718861.1 | 2962 | Missense Mutation | GAT,GTT | D,V 903 | XP_006718924.1 | |
XM_011542875.1 | 2962 | Missense Mutation | GAT,GTT | D,V 499 | XP_011541177.1 |