Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACCTTCAGGCTTCTTTCAAGTAA[C/T]TGATTAACTTCTCCATAATCATAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607768 | ||||||||||||||||||||
Literature Links: |
SESN3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SESN3 - sestrin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271594.1 | 1607 | Silent Mutation | CAA,CAG | Q,Q 288 | NP_001258523.1 | |
NM_144665.3 | 1607 | Silent Mutation | CAA,CAG | Q,Q 427 | NP_653266.2 | |
XM_011542614.1 | 1607 | Silent Mutation | CAA,CAG | Q,Q 404 | XP_011540916.1 |