Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCTCTGGTTGCAGAGCTGGCAGG[C/G]GAAGCAGTCGAGGTGATACACGTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 186921 | ||||||||||||||||||||
Literature Links: |
LMO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LMO1 - LIM domain only 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270428.1 | 527 | Missense Mutation | CCC,GCC | P,A 114 | NP_001257357.1 | |
NM_002315.2 | 527 | Missense Mutation | CCC,GCC | P,A 115 | NP_002306.1 | |
XM_006718228.3 | 527 | Missense Mutation | CCC,GCC | P,A 152 | XP_006718291.1 | |
XM_011520098.2 | 527 | Missense Mutation | CCC,GCC | P,A 104 | XP_011518400.1 | |
XM_011520099.2 | 527 | Missense Mutation | CCC,GCC | P,A 104 | XP_011518401.1 | |
XM_011520100.1 | 527 | Missense Mutation | CCC,GCC | P,A 81 | XP_011518402.1 |