Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTCAAAATGAGTTTAAAAAGGT[A/C]TCTCTCTTTTTCACAAGCTAGAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600953 | ||||||||||||||||||||
Literature Links: |
IL18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL18 - interleukin 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243211.1 | 722 | Missense Mutation | GAC,TAC | D,Y 164 | NP_001230140.1 | |
NM_001562.3 | 722 | Missense Mutation | GAC,TAC | D,Y 168 | NP_001553.1 | |
XM_011542805.1 | 722 | Missense Mutation | GAC,TAC | D,Y 164 | XP_011541107.1 | |
XM_011542806.2 | 722 | Missense Mutation | GAC,TAC | D,Y 168 | XP_011541108.1 |