Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGCACGGCCGCAGCCTCAGGGT[C/T]GAAGGCCCGAATCTCAGGCTCCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176730 MIM: 191290 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INS - insulin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
INS-IGF2 - INS-IGF2 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4686 - microRNA 4686 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TH - tyrosine hydroxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000360.3 | 3544 | Missense Mutation | AAC,GAC | N,D 413 | NP_000351.2 | |
NM_199292.2 | 3544 | Missense Mutation | AAC,GAC | N,D 444 | NP_954986.2 | |
NM_199293.2 | 3544 | Missense Mutation | AAC,GAC | N,D 440 | NP_954987.2 | |
XM_011520335.2 | 3544 | Missense Mutation | AAC,GAC | N,D 417 | XP_011518637.1 |